Novus Biologicals
Manufacturer Code:NBP2848190.1ML
Catalog # NB032747
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide corresponding to a region of Mouse DPY19L1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dpy-19-like 1; dpy-19-like 1 (C. elegans) DPY-19-like protein 1 KIAA0877; DPY-19-like protein 1; Probable C-mannosyltransferase DPY19L1; Protein dpy-19 homolog 1
Gene Aliases: DPY19L1; GA0500; KIAA0877
UniProt ID: (Human) Q2PZI1
Entrez Gene ID: (Human) 23333
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.