OriGene
Manufacturer Code:CF504366
Catalog # 50-166-2842
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D2 |
Immunogen | Full length human recombit protein of human DPP8 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Dipeptidyl peptidase 8; Dipeptidyl peptidase IV-related protein 1; dipeptidyl peptidase IV-related protein-1; Dipeptidyl peptidase VIII; dipeptidyl-peptidase 8; DP8; DP8 DPRP-1 DPRP1 MST097 MSTP097 MSTP135 MSTP141; DPP VIII; DPRP-1; Prolyl dipeptidase DPP8
Gene Aliases: 2310004I03Rik; 4932434F09Rik; AI666706; DP8; DPP VIII; DPP8; DPRP-1; DPRP1; MST097; MSTP097; MSTP135; MSTP141
UniProt ID: (Human) Q6V1X1, (Mouse) Q80YA7
Entrez Gene ID: (Dog) 487605, (Human) 54878, (Rat) 315758, (Mouse) 74388
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.