Proteintech
Manufacturer Code:127521AP150UL
Catalog # 50-172-8222
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DPP8 Fusion Protein Ag3262 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dipeptidyl peptidase 8; dipeptidyl peptidase 8 Dipeptidyl peptidase VIII DP8 DPP VIII DPP8 DPRP 1 DPRP1 MSTP135 MSTP141 Prolyl dipeptidase DPP8; Dipeptidyl peptidase IV-related protein 1; dipeptidyl peptidase IV-related protein-1; Dipeptidyl peptidase VIII; dipeptidyl-peptidase 8; DP8; DPP VIII; DPRP-1; Prolyl dipeptidase DPP8
Gene Aliases: 2310004I03Rik; 4932434F09Rik; AI666706; DP8; DPP VIII; DPP8; DPRP-1; DPRP1; MST097; MSTP097; MSTP135; MSTP141
UniProt ID: (Human) Q6V1X1, (Mouse) Q80YA7
Entrez Gene ID: (Human) 54878, (Rat) 315758, (Mouse) 74388
Molecular Function: hydrolase protease serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.