OriGene
Manufacturer Code:CF500733
Catalog # 50-165-9789
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI11D7 |
Immunogen | Full length human recombit protein of human DPP4 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ADABP; ADABP ADCP2 CD26 DPPIV TP103; ADCP-2; Adenosine deaminase complexing protein 2; CD26; Dipeptidyl peptidase 4; Dipeptidyl peptidase 4 membrane form; Dipeptidyl peptidase 4 soluble form; Dipeptidyl peptidase IV; Dipeptidyl peptidase IV membrane form; Dipeptidyl peptidase IV soluble form; dipeptidyl-peptidase 4; dipeptidylpeptidase 4; dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2); DPP IV; T-cell activation antigen CD26; TP103
Gene Aliases: ADABP; ADCP2; CD26; DPP4; DPPIV; TP103
UniProt ID: (Human) P27487
Entrez Gene ID: (Human) 1803
Molecular Function:
enzyme modulator
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.