Invitrogen
Manufacturer Code:PA521709
Catalog # PIPA521709
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 138 and 465 of Dipeptidyl-peptidase 3 (Uniprot IDNo.Q9NY33) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dipeptidyl aminopeptidase III; Dipeptidyl arylamidase III; Dipeptidyl peptidase 3; Dipeptidyl peptidase III; dipeptidyl-peptidase 3; dipeptidyl-peptidase III; dipeptidylpeptidase III; DPP III; Enkephalinase B; hCG&lowbar2016942 DPPIII DPP III dipeptidyl aminopeptidase III dipeptidyl arylamidase III dipeptidyl peptidase 3 dipeptidyl peptidase III
Gene Aliases: 4930533O14Rik; C86324; DPP3; DPPIII
UniProt ID: (Human) Q9NY33, (Mouse) Q99KK7
Entrez Gene ID: (Human) 10072, (Mouse) 75221
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.