Proteintech
Manufacturer Code:106501AP150UL
Catalog # 50-172-6480
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DPP3 Fusion Protein Ag1022 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dipeptidyl aminopeptidase III; Dipeptidyl aminopeptidase III Dipeptidyl arylamidase III dipeptidyl peptidase 3 Dipeptidyl peptidase III DPP III DPP3 DPPIII; Dipeptidyl arylamidase III; Dipeptidyl peptidase 3; Dipeptidyl peptidase III; dipeptidyl-peptidase 3; dipeptidyl-peptidase III; dipeptidylpeptidase III; DPP III; Enkephalinase B
Gene Aliases: 4930533O14Rik; C86324; DPP3; DPPIII
UniProt ID: (Human) Q9NY33, (Mouse) Q99KK7
Entrez Gene ID: (Human) 10072, (Mouse) 75221
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.