Invitrogen
Manufacturer Code:MA525706
Catalog # PIMA525706
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2A5 |
Immunogen | Full length human recombit protein of DPP10 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: dipeptidyl peptidase 10; Dipeptidyl peptidase IV-related protein 3; Dipeptidyl peptidase X; Dipeptidyl peptidase-like protein 2; dipeptidyl-peptidase 10 (inactive); dipeptidyl-peptidase 10 (non-functional); DPL2; DPP X; DPP10 dipeptidylpeptidase 10 HGNC:20823 DPRP3 dipeptidyl peptidase IV-related protein 3 dipeptidyl peptidase X dipeptidyl peptidase-like protein 2 dipeptidyl-peptidase 10 DPL2 DPPY OTTHUMP00000203618 dipeptidyl peptidase 10 dipeptidyl peptidase like protein 2; DPRP-3; Inactive dipeptidyl peptidase 10
Gene Aliases: DPL2; DPP10; DPPY; DPRP-3; DPRP3; KIAA1492
UniProt ID: (Human) Q8N608
Entrez Gene ID: (Human) 57628
Molecular Function: enzyme modulator hydrolase protease serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.