OriGene
Manufacturer Code:TA503640
Catalog # 50-167-4484
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2A5 |
Immunogen | Full length human recombit protein of human DPP10 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: dipeptidyl peptidase 10; Dipeptidyl peptidase IV-related protein 3; Dipeptidyl peptidase X; Dipeptidyl peptidase-like protein 2; dipeptidyl-peptidase 10 (inactive); dipeptidyl-peptidase 10 (non-functional); DPL2; DPL2 DPPY DPRP-3 DPRP3; DPP X; DPRP-3; Inactive dipeptidyl peptidase 10
Gene Aliases: DPL2; DPP10; DPPY; DPRP-3; DPRP3; KIAA1492
UniProt ID: (Human) Q8N608
Entrez Gene ID: (Human) 57628
Molecular Function: enzyme modulator hydrolase protease serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.