Invitrogen
Manufacturer Code:PA537949
Catalog # PIPA537949
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence HDCEDAARELQSQ from the internal region of DPM3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Dolichol-phosphate mannose synthase subunit 3; Dolichol-phosphate mannosyltransferase subunit 3; Dolichyl-phosphate beta-D-mannosyltransferase subunit 3; dolichyl-phosphate mannosyltransferase polypeptide 3; dolichyl-phosphate mannosyltransferase polypeptide 3 MGC125904 MGC125905 MGC34275 dolichol-phosphate mannose synthase subunit 3 dolichyl-phosphate beta-D-mannosyltransferase subunit 3 mannose-P-dolichol synthase subunit 3 prostin 1; DPM synthase complex subunit 3; DPM synthase subunit 3; Mannose-P-dolichol synthase subunit 3; MPD synthase subunit 3; prostin 1; Prostin-1; testicular tissue protein Li 58
Gene Aliases: CDG1O; DPM3
UniProt ID: (Human) Q9P2X0
Entrez Gene ID: (Human) 54344
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.