Novus Biologicals
Manufacturer Code:NBP238056
Catalog # NBP238056
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: SSPPPLDPGQGFGKTPPLNMLPHFGGAGSCLPRTHPWGFPVHPSRLPVSNLWVSWC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dolichol phosphate-mannose biosynthesis regulatory protein; dolichol phosphate-mannose biosynthesis regulatory protein dolichol phosphate-mannose synthase 2 dolichyl-phosphate mannosyltransferase polypeptide 2 regulatory subunit FLJ80013 MGC111193 MGC21559; Dolichol-phosphate mannose synthase subunit 2; DPM synthase complex subunit; DPM synthase subunit 2
Gene Aliases: CDG1U; DPM2; My026
UniProt ID: (Human) O94777
Entrez Gene ID: (Human) 8818
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.