Abnova
Manufacturer Code:MAB14032
Catalog # 89-035-437
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5C5A7 |
Immunogen | Yeast Dolichol phosphate mannose synthase. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Dolichol-phosphate mannose synthase; Dolichol-phosphate mannosyltransferase; Dolichyl-phosphate beta-D-mannosyltransferase; DPM synthase; Mannose-P-dolichol synthase; MPD synthase; SED3
Gene Aliases: DPM1; P9705.3; SED3; YPR183W
UniProt ID: (Baker's yeast) P14020
Entrez Gene ID: (Baker's yeast) 856313
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.