Invitrogen
Manufacturer Code:PA5112404
Catalog # PIPA5112404
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Dolichol-phosphate mannosyltransferase subunit 1 protein (2-260AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI118379 AI194292 CDGIE dolichol monophosphate mannose synthase dolichol-phosphate (beta-D) mannosyltransferase 1 Dolichol-phosphate mannose synthase Dolichol-phosphate mannose synthase subunit 1 Dolichol-phosphate mannosyltransferase Dolichol-phosphate mannosyltransferase subunit 1 Dolichyl-phosphate beta-D-mannosyltransferase Dolichyl-phosphate beta-D-mannosyltransferase subunit 1 dolichyl-phosphate mannosyltransferase 1 dolichyl-phosphate mannosyltransferase polypeptide 1 catalytic subunit dolichyl-phosphate mannosyltransferase polypeptide 1 catalytic subunit DPM synthase DPM synthase complex catalytic subunit DPM synthase subunit 1 DPM1 Mannose-P-dolichol synthase Mannose-P-dolichol synthase subunit 1 MPD synthase MPD synthase subunit 1 MPDS OTTMUSP00000017164 P9705.3 RP23-391M18.1 SED3 YPR183W; dolichol monophosphate mannose synthase; Dolichol-phosphate mannose synthase subunit 1; Dolichol-phosphate mannosyltransferase subunit 1; Dolichyl-phosphate beta-D-mannosyltransferase subunit 1; dolichyl-phosphate mannosyltransferase polypeptide 1 catalytic subunit; DPM synthase complex, catalytic subunit; DPM synthase subunit 1; Mannose-P-dolichol synthase subunit 1; MPD synthase; MPD synthase subunit 1
Gene Aliases: AI118379; AI194292; CDGIE; DPM1; MPDS
UniProt ID: (Human) O60762, (Mouse) O70152
Entrez Gene ID: (Human) 8813, (Mouse) 13480
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.