Invitrogen
Manufacturer Code:PA530226
Catalog # PIPA530226
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 25 and 286 of Human DPH2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-(3-amino-3-carboxypropyl)histidine synthase subunit 2; Diphthamide biosynthesis protein 2; diphthamide biosynthesis protein 2 homolog-like 2; diphthamide biosynthesis-like protein 2; Diphtheria toxin resistance protein 2; diptheria toxin resistance protein required for diphthamide biosynthesis-like 2; DPH2-like 2; DPH2L2 DPH2-like 2 diphthamide biosynthesis protein 2 diphthamide biosynthesis protein 2 homolog-like 2 diphthamide biosynthesis-like protein 2 diptheria toxin resistance protein required for diphthamide biosynthesis-like 2; S-adenosyl-L-methionine:L-histidine 3-amino-3-carboxypropyltransferase 2
Gene Aliases: DPH2; DPH2L2
UniProt ID: (Human) Q9BQC3
Entrez Gene ID: (Human) 1802
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.