Proteintech
Manufacturer Code:122221AP150UL
Catalog # 50-172-7808
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DPEP1 Fusion Protein Ag2907 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-lactamase; Dehydropeptidase I Dipeptidase 1 dipeptidase 1 (renal) DPEP1 hRDP MBD1 MDP Microsomal dipeptidase RDP Renal dipeptidase; Dehydropeptidase-I; Dipeptidase 1; DPEP-1; hRDP; MBD-1; membrane bound dipeptidase type II; Membrane-bound dipeptidase 1; Microsomal dipeptidase; Renal dipeptidase; testicular tissue protein Li 57
Gene Aliases: AI327012; DPEP1; MBD; MBD1; MDP; RDP
UniProt ID: (Human) P16444, (Rat) P31430, (Mouse) P31428
Entrez Gene ID: (Human) 1800, (Rat) 94199, (Mouse) 13479
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.