Invitrogen
Manufacturer Code:PA526825
Catalog # PIPA526825
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 96-136 amino acids from the central region of human DPCD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: deleted in a mouse model of primary ciliary dyskinesia; deleted in a mouse model of primary ciliary dyskinesia protein DPCD RP11-529I104; neighbor of Dac; Protein DPCD; RP11-529I10.4
Gene Aliases: 5330431N19Rik; DPCD; Ndac
UniProt ID: (Human) Q9BVM2, (Mouse) Q8BPA8
Entrez Gene ID: (Human) 25911, (Mouse) 226162
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.