Invitrogen
Manufacturer Code:PA545785
Catalog # PIPA545785
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human DPAGT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dolichyl-phosphate (UDP-N-acetylglucosamine) acetylglucosaminephosphotransferase 1 (GlcNAc-1-P transferase); dolichyl-phosphate (UDP-N-acetylglucosamine) N-acetylglucosaminephosphotransferase 1 (GlcNAc-1-P tra; dolichyl-phosphate (UDP-N-acetylglucosamine) N-acetylglucosaminephosphotransferase 1 (GlcNAc-1-P transferase); dolichyl-phosphate (UDP-N-acetylglucosamine) N-acetylglucosaminephosphotransferase 1 (GlcNAc-1-P transferase) ALG7 CDG-Ij D11S366 DGPT DPAGT DPAGT2 G1PT GPT UAGT UGAT CDG1J; dolichyl-phosphate alpha-N-acetylglucosaminephosphotransferase 2; dolichyl-phosphate alpha-N-acetylglucosaminyltransferase; G1PT; GlcNAc-1-P transferase; GlcNAc-1-P transferase 1; N-acetylglucosamine-1-phosphate transferase; UDP-GlcNAc:dolichyl-phosphate N-acetylglucosaminephosphotransferase; UDP-N-acetylglucosamine lysosomal enzyme N-acetylglucosamine phosphotransferase; UDP-N-acetylglucosamine--dolichyl-phosphate N-acetylglucosaminephosphotransferase; UDP-N-acetylglucosamine--dolichyl-phosphate N-acetylglucosaminephosphotransferase 1; UDP-N-acetylglucosamine-dolichyl-phosphate N-acetylglucosaminephosphotransferase
Gene Aliases: ALG7; AU021132; CDG-Ij; CDG1J; CMS13; CMSTA2; D11S366; DGPT; DPAGT; DPAGT1; DPAGT2; G1PT; Gnpta; GPT; H2afx; UAGT; UGAT
UniProt ID: (Human) Q9H3H5, (Mouse) P42867
Entrez Gene ID: (Human) 1798, (Dog) 489371, (Rabbit) 100359180, (Horse) 100063303, (Guinea pig) 100720260, (Rat) 300668, (Mouse) 13478
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.