Proteintech
Manufacturer Code:110152AP150UL
Catalog # 50-172-6797
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DOM3Z Fusion Protein Ag1491 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5'-3' exoribonuclease DXO; Decapping and exoribonuclease protein; DeNADding enzyme DXO; Dom 3 homolog Z dom 3 homolog Z (C. elegans) DOM3L DOM3Z DXO NG6 Protein Dom3Z; Dom-3 homolog Z; DXO; NAD-capped RNA hydrolase DXO; protein Dom3Z
Gene Aliases: DOM3L; DOM3Z; DXO; NG6; RAI1
UniProt ID: (Human) Q5ST60, (Rat) Q6MG77, (Mouse) O70348
Entrez Gene ID: (Human) 1797, (Rat) 361799, (Mouse) 112403
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.