Invitrogen
Manufacturer Code:PA565874
Catalog # PIPA565874
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human DOK5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Docking protein 5; DOK5 MGC16926 C20orf180 docking protein 5 downstream of tyrosine kinase 5 chromosome 20 open reading frame 180; Downstream of tyrosine kinase 5; Insulin receptor substrate 6; IRS-6
Gene Aliases: C20orf180; DOK5; IRS-6; IRS6
UniProt ID: (Human) Q9P104
Entrez Gene ID: (Human) 55816
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.