Invitrogen
Manufacturer Code:PA514757
Catalog # PIPA514757
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 276-306 amino acids from the C-terminal region of human DOK5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Docking protein 5; Downstream of tyrosine kinase 5; Downstream of tyrosine kinase 5 Protein dok-5; Insulin receptor substrate 6; IRS-6
Gene Aliases: 2700055C10Rik; C20orf180; DOK5; IRS-6; IRS6
UniProt ID: (Human) Q9P104, (Mouse) Q91ZM9
Entrez Gene ID: (Human) 55816, (Mouse) 76829
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.