Novus Biologicals
Manufacturer Code:NBP239051
Catalog # NBP239051
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: EQHERLLQSVKNSMLQMKMSERAASLSTMVPLPRSAY |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C20orf180 chromosome 20 open reading frame 180 dJ805C22.1 docking protein 5 Downstream of tyrosine kinase 5 Insulin receptor substrate 6 IRS6 IRS-6 MGC16926; Docking protein 5; Downstream of tyrosine kinase 5; Insulin receptor substrate 6; IRS-6
Gene Aliases: C20orf180; DOK5; IRS-6; IRS6
UniProt ID: (Human) Q9P104
Entrez Gene ID: (Human) 55816
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.