Proteintech
Manufacturer Code:189871AP150UL
Catalog # 50-173-2307
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cdc42 guanine nucleotide exchange factor zizimin-1; dedicator of cytokinesis 9 DOCK9 KIAA1058 KIAA1085 ZIZ1 ZIZIMIN1; Dedicator of cytokinesis protein 9; Zizimin-1
Gene Aliases: AA959601; AW538057; B230309H04Rik; D14Wsu89e; DOCK9; KIAA1058; mKIAA1058; ZIZ1; ZIZIMIN1
UniProt ID: (Human) Q9BZ29
Entrez Gene ID: (Human) 23348, (Mouse) 105445
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.