OriGene
Manufacturer Code:CF801863
Catalog # 50-166-5748
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI12D7 |
Immunogen | Human recombit protein fragment corresponding to amino acids 84-405 of human DOCK8 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 1200017A24Rik; Dedicator of cytokinesis protein 8; epididymis luminal protein 205; HEL-205 MRD2 ZIR8
Gene Aliases: DOCK8; HEL-205; MRD2; ZIR8
UniProt ID: (Human) Q8NF50
Entrez Gene ID: (Human) 81704
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.