Invitrogen
Manufacturer Code:PA5103779
Catalog # PIPA5103779
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DOCK7 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3110056M06Rik dedicator of cytokinesis 7 dedicator of cytokinesis protein 7 DOCK7 EIEE23 Gm430 KIAA1771 m protein moonlight ZIR2; Dedicator of cytokinesis protein 7; protein moonlight
Gene Aliases: 3110056M06Rik; DOCK7; EIEE23; Gm430; KIAA1771; m; ZIR2
UniProt ID: (Human) Q96N67
Entrez Gene ID: (Human) 85440, (Mouse) 67299, (Rat) 313388
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.