Novus Biologicals
Manufacturer Code:NBP178749
Catalog # NBP178749
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this product maps to a region between residue 2090 and 2140 of human Dedicator of Cytokinesis 7 using the numbering given in entry Q96N67.4 (GeneID 85440). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dedicator of cytokinesis 7 dedicator of cytokinesis protein 7 KIAA1771ZIR2; Dedicator of cytokinesis protein 7
Gene Aliases: DOCK7; EIEE23; KIAA1771; ZIR2
UniProt ID: (Human) Q96N67
Entrez Gene ID: (Human) 85440
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.