Novus Biologicals
Manufacturer Code:NBP257555
Catalog # NBP257555
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:CSAIYPTPVEPSQRMLFNHIGDGALPRSDPNLSAPEKAVNPTPSSWSLDSGKEAKNMSDSGKLISP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dedicator of cytokinesis 4 dedicator of cytokinesis protein 4 KIAA0716FLJ34238 MGC134911 MGC134912; Dedicator of cytokinesis protein 4
Gene Aliases: DOCK4; KIAA0716
UniProt ID: (Human) Q8N1I0
Entrez Gene ID: (Human) 9732
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.