Invitrogen
Manufacturer Code:PA576783
Catalog # PIPA576783
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human DNase I. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deoxyribonuclease 1 Deoxyribonuclease I Deoxyribonuclease-1 Deoxyribonuclease1 DeoxyribonucleaseI DNAS1&lowbarHUMAN DNASE 1 DNase I DNase I lysosomal DNASE1 DNaseI DNL 1 DNL1 Dornase alfa DRNI FLJ38093 Human urine deoxyribonuclease I; Deoxyribonuclease I; Deoxyribonuclease-1; DNase I; DNase I, lysosomal; Dornase alfa; human urine deoxyribonuclease I
Gene Aliases: AI788650; DNASE1; DNaseI; DNL1; DRNI
UniProt ID: (Human) P24855, (Mouse) P49183
Entrez Gene ID: (Human) 1773, (Mouse) 13419
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.