Proteintech
Manufacturer Code:192211AP150UL
Catalog # 50-173-2370
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DNMT2 Fusion Protein Ag5571 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA (cytosine-5)-methyltransferase-like protein 2; DNA cytosine-5 methyltransferase 2; DNA methyltransferase 2; DNA methyltransferase homolog HsaIIP; DNA methyltransferase homolog MmuIIP; DNA methyltransferase-2; DNA MTase homolog HsaIIP; DNA MTase homolog HsaIIP DNMT2 M.HsaIIP PuMet RNMT1 TRDMT1; DNA MTase homolog MmuIIP; Dnmt2; m.MmuIIP; Met-2; PuMet; tRNA (cytosine(38)-C(5))-methyltransferase; tRNA (cytosine-5-)-methyltransferase
Gene Aliases: DMNT2; DNMT2; Met2; MHSAIIP; PUMET; RNMT1; Rnmt2; TRDMT1
UniProt ID: (Human) O14717, (Mouse) O55055, (Rat) Q4G073
Entrez Gene ID: (Human) 1787, (Mouse) 13434, (Rat) 291324
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.