Novus Biologicals
Manufacturer Code:NB100264
Catalog # NB100264
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Contained within amino acids 1-125 of the N-terminus of human Dnmt1. [UniProt# P26358] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AIM CXXC9CXXC finger protein 9 CXXC-type zinc finger protein 9 DNA (cytosine-5-)-methyltransferase 1 DNA (cytosine-5)-methyltransferase 1 DNA methyltransferase 1 DNA methyltransferase HsaI DNA MTase HsaI DNMT Dnmt1 EC 2.1.1.37 FLJ16293 m.HsaI MCMTMGC104992; CXXC-type zinc finger protein 9; DNA (cytosine-5)-methyltransferase 1; DNA methyltransferase HsaI; DNA MTase HsaI; Dnmt1; MCMT
Gene Aliases: ADCADN; AIM; CXXC9; DNMT; DNMT1; HSN1E; m.HsaI; MCMT
UniProt ID: (Human) P26358
Entrez Gene ID: (Human) 1786
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.