R&D Systems
Manufacturer Code:AF6110SP
Catalog # AF6110SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human DNMT1 Ala1381-Ala1587 Accession # P26358 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: AIM CXXC9 CXXC9CXXC finger protein 9 CXXC-type zinc finger protein 9 DNA (cytosine-5-)-methyltransferase 1 DNA (cytosine-5)-methyltransferase 1 DNA methyltransferase 1 DNA methyltransferase HsaI DNA MTase HsaI DNMT Dnmt1 EC 2.1.1.37 FLJ16293 m.HsaI MCMT MCMTMGC104992; CXXC-type zinc finger protein 9; DNA (cytosine-5)-methyltransferase 1; DNA methyltransferase HsaI; DNA MTase HsaI; Dnmt1; MCMT
Gene Aliases: ADCADN; AIM; CXXC9; DNMT; DNMT1; HSN1E; m.HsaI; MCMT
UniProt ID: (Human) P26358
Entrez Gene ID: (Human) 1786
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.