Proteintech
Manufacturer Code:146051AP150UL
Catalog # 50-172-9578
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DNM2 Fusion Protein Ag6156 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CMTDI1 CMTDIB DI CMTB DNM2 DYN2 dynamin 2 DYNII; dynamin II; Dynamin UDNM; Dynamin-2
Gene Aliases: b2b2159Clo; CMT2M; CMTDI1; CMTDIB; DI-CMTB; DNM2; DYIIAAB; DYN2; DYNII; LCCS5; Udnm
UniProt ID: (Human) P50570, (Rat) P39052, (Mouse) P39054
Entrez Gene ID: (Human) 1785, (Rat) 25751, (Mouse) 13430
Molecular Function:
G-protein
cytoskeletal protein
enzyme modulator
hydrolase
microtubule family cytoskeletal protein
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.