Novus Biologicals
Manufacturer Code:NBP216198
Catalog # NBP216198
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human DNase IL3. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deoxyribonuclease gamma; deoxyribonuclease gamma deoxyribonuclease I-like 3Liver and spleen DNase deoxyribonuclease I-like III DNAS1L3LSDDHP2 DNase gamma DNase I homolog protein 2 DNase I homolog protein DHP2 DNase I-like 3 EC 3.1.21.- EC 3.1.21.1 LS-DNase; Deoxyribonuclease I-like 3; deoxyribonuclease I-like III; DNase gamma; DNase I homolog protein 2; DNase I homolog protein DHP2; DNase I-like 3; Liver and spleen DNase; LS-DNase
Gene Aliases: DHP2; DNAS1L3; DNASE1L3; LSD; SLEB16
UniProt ID: (Human) Q13609
Entrez Gene ID: (Human) 1776
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.