Proteintech
Manufacturer Code:113301AP150UL
Catalog # 50-172-7066
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DNASE1L3 Fusion Protein Ag1873 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deoxyribonuclease gamma; Deoxyribonuclease gamma deoxyribonuclease I like 3 DHP2 DNAS1L3 DNase gamma DNase I homolog protein DHP2 DNase I like 3 DNASE1L3 Liver and spleen DNase LS DNase LSD; Deoxyribonuclease I-like 3; deoxyribonuclease I-like III; DNase gamma; DNase I homolog protein 2; DNase I homolog protein DHP2; DNase I-like 3; DNaseY; Liver and spleen DNase; LS-DNase; macrophage specific DNase I
Gene Aliases: DHP2; DNAS1L3; DNASE1L3; DNasegamma; LSD; SLEB16
UniProt ID: (Human) Q13609, (Mouse) O55070, (Rat) O89107
Entrez Gene ID: (Human) 1776, (Mouse) 13421, (Rat) 116687
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.