Novus Biologicals
Manufacturer Code:NBP212268G
Catalog # NBP212268G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0248-1X20 |
Immunogen | Human CD226 |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: adhesion glycoprotein; CD226; CD226 antigen; CD226 antigenplatelet and T cell activation antigen 1 CD226 molecule DNAM1adhesion glycoprotein DNAM-1DNAX accessory molecule-1 DNAX accessory molecule 1 PTA1 T lineage-specific activation antigen 1 antigen TLiSA1; DNAM-1; DNAX accessory molecule 1; DNAX accessory molecule-1; platelet and T cell activation antigen 1; T lineage-specific activation antigen 1 antigen
Gene Aliases: CD226; DNAM-1; DNAM1; PTA1; TLiSA1
UniProt ID: (Human) Q15762
Entrez Gene ID: (Human) 10666
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.