R&D Systems
Manufacturer Code:FAB666A
Catalog # FAB666A
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 102511 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human DNAM-1/CD226 Glu19-Asn247 (predicted) Accession # Q15762 |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: adhesion glycoprotein; CD226; CD226 antigen; CD226 CD226 antigenplatelet and T cell activation antigen 1 CD226 molecule DNAM1 DNAM1adhesion glycoprotein DNAM-1DNAX accessory molecule-1 DNAX accessory molecule 1 PTA1 T lineage-specific activation antigen 1 antigen TLiSA1; DNAM-1; DNAX accessory molecule 1; DNAX accessory molecule-1; platelet and T cell activation antigen 1; T lineage-specific activation antigen 1 antigen
Gene Aliases: CD226; DNAM-1; DNAM1; PTA1; TLiSA1
UniProt ID: (Human) Q15762
Entrez Gene ID: (Human) 10666
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.