Invitrogen
Manufacturer Code:PA530197
Catalog # PIPA530197
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 31 and 244 of Human DNAJC7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DJ11 DJC7 TPR2 TTC2 TPR repeat protein 2 dnaJ homolog subfamily C member 7 tetratricopeptide repeat domain 2 tetratricopeptide repeat protein 2; DnaJ (Hsp40) homolog, subfamily C, member 7; DnaJ homolog subfamily C member 7; tetratricopeptide repeat domain 2; Tetratricopeptide repeat protein 2; TPR repeat protein 2
Gene Aliases: DJ11; DJC7; DNAJC7; TPR2; TTC2
UniProt ID: (Human) Q99615
Entrez Gene ID: (Human) 7266
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.