Invitrogen
Manufacturer Code:PA5115135
Catalog # PIPA5115135
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DNAJC7 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2010003F24Rik 2010004G07Rik CCRP Cytoplasmic CAR retention protein cytoplasmic constitutive active/androstane receptor retention protein DJ11 DJC7 DnaJ (Hsp40) homolog subfamily C member 7 DnaJ heat shock protein family (Hsp40) member C7 dnaJ homolog subfamily C member 7 Dnajc7 MDj11 mTpr2 tetratricopeptide repeat domain 2 tetratricopeptide repeat protein 2 TPR repeat protein 2 TPR2 TTC2; CCRP; Cytoplasmic CAR retention protein; DnaJ (Hsp40) homolog, subfamily C, member 7; DnaJ homolog subfamily C member 7; MDj11; tetratricopeptide repeat domain 2; Tetratricopeptide repeat protein 2; TPR repeat protein 2
Gene Aliases: 2010003F24Rik; 2010004G07Rik; CCRP; DJ11; DJC7; DNAJC7; mDj11; mTpr2; TPR2; TTC2
UniProt ID: (Human) Q99615, (Mouse) Q9QYI3
Entrez Gene ID: (Human) 7266, (Mouse) 56354
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.