Novus Biologicals
Manufacturer Code:NBP287291
Catalog # NB034913
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human DNAJC6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: auxilin; DJC6 DnaJ (Hsp40) homolog subfamily B member 6 DnaJ (Hsp40) homolog subfamily C member 6 DnaJ homolog subfamily C member 6 EC 3.1.3.48 KIAA0473auxilin MGC129914 MGC129915 MGC48436 putative tyrosine-protein phosphatase auxilin; DnaJ (Hsp40) homolog, subfamily B, member 6; DnaJ (Hsp40) homolog, subfamily C, member 6; DnaJ homolog subfamily C member 6; Putative tyrosine-protein phosphatase auxilin
Gene Aliases: DJC6; DNAJC6; KIAA0473; PARK19
UniProt ID: (Human) O75061
Entrez Gene ID: (Human) 9829
Molecular Function: chaperone kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.