Proteintech
Manufacturer Code:219411AP150UL
Catalog # 50-173-3652
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DNAJC6/AUXILIN Fusion Protein Ag16612 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: auxilin; AUXILIN DJC6 DNAJC6 KIAA0473; DnaJ (Hsp40) homolog, subfamily B, member 6; DnaJ (Hsp40) homolog, subfamily C, member 6; DnaJ homolog subfamily C member 6; Putative tyrosine-protein phosphatase auxilin
Gene Aliases: 2810027M23Rik; DJC6; DNAJC6; KIAA0473; mKIAA0473; PARK19
UniProt ID: (Human) O75061, (Mouse) Q80TZ3
Entrez Gene ID: (Human) 9829, (Mouse) 72685, (Rat) 313409
Molecular Function: chaperone kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.