Proteintech
Manufacturer Code:160631AP150UL
Catalog # 50-173-0686
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DNAJC15 Fusion Protein Ag8911 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell growth-inhibiting gene 22 protein; DnaJ (Hsp40) homolog, subfamily C, member 15; DnaJ (Hsp40) homolog, subfamily D, member 1; DNAJ domain-containing; DnaJ homolog subfamily C member 15; DNAJC15 DNAJD1 HSD18 MCJ; MCJ; Methylation-controlled J protein
Gene Aliases: 1110003P16Rik; DNAJC15; DNAJD1; GIG22; HSD18; MCJ
UniProt ID: (Human) Q9Y5T4, (Mouse) Q78YY6
Entrez Gene ID: (Human) 29103, (Rat) 290370, (Mouse) 66148
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.