Invitrogen
Manufacturer Code:PA543346
Catalog # PIPA543346
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human DNAJC10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DnaJ (Hsp40) homolog, subfamily C, member 10; DnaJ homolog subfamily C member 10; DnaJ-like protein subfamily C member 10-like protein; Endoplasmic reticulum DNA J domain-containing protein 5; Endoplasmic reticulum DnaJ-PDI fusion protein 1; ER-resident protein ERdj5; J domain-containing PDI-like protein; J domain-containing protein disulfide isomerase-like protein; J-domain-containing protein disulfide isomerase-like protein; Macrothioredoxin; MTHr; protein disulfide isomerase family A, member 19
Gene Aliases: 1200006L06Rik; D2Ertd706e; DNAJC10; ERDJ5; JPDI; MTHr; PDIA19; UNQ495/PRO1012; zgc:162218
UniProt ID: (Human) Q8IXB1, (Rat) Q498R3, (Mouse) Q9DC23
Entrez Gene ID: (Dog) 478826, (Horse) 102147457, (Human) 54431, (Bovine) 613403, (Rabbit) 100359290, (Guinea pig) 100716408, (Rat) 295690, (Mouse) 66861, (Zebrafish) 557858
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.