Novus Biologicals
Manufacturer Code:NBP270951PEATT594
Catalog # N270951P594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4D12 |
Immunogen | Full length recombinant protein of human DNAJB2 (NP_001034639) produced in HEK293T cells. |
Conjugate | PE/ATTO 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Mouse Monoclonal Antibody
Protein Aliases: DnaJ (Hsp40) homolog subfamily B member 2 HSJ-1 HSPF3neuronal DNAJ-like 1; DnaJ (Hsp40) homolog, subfamily B, member 2; DnaJ homolog subfamily B member 2; dnaJ protein homolog 1; Heat shock 40 kDa protein 3; Heat shock protein J1; heat shock protein, neuronal DNAJ-like 1; HSJ-1
Gene Aliases: CMT2T; DNAJB2; DSMA5; HSJ-1; HSJ1; HSPF3
UniProt ID: (Human) P25686
Entrez Gene ID: (Human) 3300
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.