Invitrogen
Manufacturer Code:PA599813
Catalog # PIPA599813
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DNAJB2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2700059H22Rik CMT2T DnaJ (Hsp40) homolog subfamily B member 10 DnaJ (Hsp40) homolog subfamily B member 2 DnaJ heat shock protein family (Hsp40) member B2 dnaJ homolog subfamily B member 10 DnaJ homolog subfamily B member 2 dnaJ protein homolog 1 Dnajb10 DNAJB2 DSMA5 heat shock 40 kDa protein 3 Heat shock protein J1 heat shock protein DNAJ-like 1 heat shock protein neuronal DNAJ-like 1 HSJ1 HSJ-1 HSPF3 mDj8; DnaJ (Hsp40) homolog, subfamily B, member 2; DnaJ homolog subfamily B member 2; dnaJ protein homolog 1; Heat shock 40 kDa protein 3; Heat shock protein J1; heat shock protein, neuronal DNAJ-like 1; HSJ-1
Gene Aliases: CMT2T; DNAJB2; DSMA5; HSJ-1; HSJ1; HSPF3
UniProt ID: (Human) P25686
Entrez Gene ID: (Human) 3300
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.