Proteintech
Manufacturer Code:108381AP150UL
Catalog # 50-172-6643
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DNAJB2 Fusion Protein Ag1271 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DnaJ (Hsp40) homolog, subfamily B, member 10; DnaJ (Hsp40) homolog, subfamily B, member 2; DnaJ homolog subfamily B member 10; DnaJ homolog subfamily B member 2; dnaJ protein homolog 1; DnaJ protein homolog 1 DNAJB2 Heat shock 40 kDa protein 3 Heat shock protein J1 HSJ 1 HSJ1 HSPF3; Heat shock 40 kDa protein 3; Heat shock protein J1; heat shock protein, DNAJ-like 1; heat shock protein, neuronal DNAJ-like 1; HSJ-1; mDj8
Gene Aliases: 2700059H22Rik; CMT2T; Dnajb10; DNAJB2; DSMA5; HSJ-1; HSJ1; HSPF3; mDj8
UniProt ID: (Human) P25686, (Mouse) Q9QYI5
Entrez Gene ID: (Human) 3300, (Mouse) 56812, (Rat) 689593
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.