OriGene
Manufacturer Code:CF502244
Catalog # 50-166-1168
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1G4 |
Immunogen | Full length human recombit protein of human DNAJB1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DnaJ (Hsp40) homolog, subfamily B, member 1; DnaJ homolog subfamily B member 1; DnaJ protein homolog 1; hDj-1; Heat shock 40 kDa protein 1; HSP40; HSPF1 Hdj1 Hsp40 RSPH16B Sis1; Human DnaJ protein 1; radial spoke 16 homolog B
Gene Aliases: DNAJ1; DNAJB1; HDJ1; Hsp40; HSPF1; RSPH16B; Sis1
UniProt ID: (Human) P25685
Entrez Gene ID: (Human) 3337, (Dog) 476688
Molecular Function:
chaperone
chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.