Proteintech
Manufacturer Code:175331AP150UL
Catalog # 50-173-1629
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DNAI2 Fusion Protein Ag11585 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Axonemal dynein intermediate chain 2; CILD9 DNAI2; Dynein intermediate chain 2, axonemal; dynein, axonemal, intermediate chain 2; dynein, axonemal, intermediate polypeptide 2
Gene Aliases: C030015H18Rik; CILD9; DIC2; DNAI2; Dnaic2; Gm728
UniProt ID: (Human) Q9GZS0, (Rat) Q66HC9, (Mouse) A2AC93
Entrez Gene ID: (Human) 64446, (Rat) 360654, (Mouse) 432611
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.