Abnova
Manufacturer Code:PAB23062
Catalog # 89-124-762
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human DNAH1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant DNAH1.
For Research Use Only
Protein Aliases: Axonemal beta dynein heavy chain 1; Ciliary dynein heavy chain 1; DNAH1 variant protein; DNAHC1; Dynein heavy chain 1, axonemal; dynein, axonemal, heavy chain 1; dynein, axonemal, heavy polypeptide 1; HDHC7; Heat shock regulated protein 1; HL-11; HL11; HSRF-1; testicular tissue protein Li 60; XLHSRF-1
Gene Aliases: DHC7; DNAH1; DNAHC1; HDHC7; HL-11; HL11; HSRF-1; KIAA1410; XLHSRF-1
UniProt ID: (Human) Q9P2D7
Entrez Gene ID: (Human) 25981
Molecular Function:
cytoskeletal protein
hydrolase
microtubule binding motor protein
microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.