Invitrogen
Manufacturer Code:PA544697
Catalog # PIPA544697
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human C14orf104 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dynein (axonemal) assembly factor 2; Dynein assembly factor 2, axonemal; dynein, axonemal, assembly factor 2; kintoun; kintoun homolog; Protein kintoun
Gene Aliases: 1110034A24Rik; 2810020C19Rik; AV032410; C10H14orf104; C14orf104; CILD10; DNAAF2; kintoun; KTU; mknt; PF13; RGD1310311; TU54
UniProt ID: (Bovine) Q0VC73, (Human) Q9NVR5, (Rat) Q5FVL7, (Mouse) Q8BPI1
Entrez Gene ID: (Bovine) 534465, (Dog) 480313, (Horse) 100065764, (Pig) 100521173, (Human) 55172, (Rat) 362746, (Mouse) 109065
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.