Proteintech
Manufacturer Code:215991AP150UL
Catalog # 50-173-3505
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DNA2 Fusion Protein Ag16206 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA replication ATP-dependent helicase DNA2; DNA replication ATP-dependent helicase-like homolog; DNA replication ATP-dependent helicase/nuclease DNA2; DNA replication helicase 2 homolog; DNA replication nuclease DNA2; DNA2 DNA2L KIAA0083; DNA2-like helicase; hDNA2
Gene Aliases: DNA2; DNA2L; hDNA2; KIAA0083
UniProt ID: (Human) P51530
Entrez Gene ID: (Human) 1763
Molecular Function:
DNA binding protein
DNA helicase
RNA binding protein
RNA helicase
endoribonuclease
helicase
hydrolase
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.