Novus Biologicals
Manufacturer Code:NBP136950
Catalog # NBP136950
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence DTHQQLTCSNDES corresponding to internal region according to NP_955452.3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp781A0112 DNA polymerase eta EC 2.7.7.7 POLHDNA polymerase theta polymerase (DNA directed) theta PRO0327; DNA polymerase eta; DNA polymerase theta; polymerase (DNA directed), theta; polymerase (DNA) theta
Gene Aliases: POLH; POLQ; PRO0327
UniProt ID: (Human) O75417
Entrez Gene ID: (Human) 10721
Molecular Function:
DNA binding protein
DNA-directed DNA polymerase
exodeoxyribonuclease
hydrolase
nuclease
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.