Novus Biologicals
Manufacturer Code:NBP292047100UL
Catalog # NB025986
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-170 of human PRIM2 (NP_000938.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dJ422B11.1.1 DNA primase 58 kDa subunit DNA primase subunit p58 EC 2.7.7 EC 2.7.7.- MGC75142 p58 polypeptide 2A p58 PRIM2ADNA primase large subunit primase polypeptide 2A 58kDa primase DNA polypeptide 2 (58kDa) primase polypeptide 2A (58kD) primase polypeptide 2A 58kDa; DNA primase 58 kDa subunit; DNA primase large subunit; p58; primase, DNA, polypeptide 2 (58kDa)
Gene Aliases: p58; PRIM2; PRIM2A
UniProt ID: (Human) P49643
Entrez Gene ID: (Human) 5558
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.